Brady™ Indoor/Outdoor Vinyl Label Cartridges
Adheres to irregular, rough and curved surfaces
Manufacturer:  Brady™ M21375595BR
 View more versions of this product
Catalog No. 19-102-813
Description
Description
Adheres to irregular, rough and curved surfaces
  • For use with BMP™21, BMP21-LAB, BMP21-PLUS, IDPal™, LabPal™ printers
  • Made of 8-10 year indoor/outdoor material
  • Matte finish with permanent adhesive
  • Stands up to grease, oil and most industrial chemicals
  • Available in various sizes and colors
Applications:
General identification, laboratory identification, panel identification, panel labels, plant labels, safety labels, Lean/5S and visual workplace labels, pipe marking, tool labeling, equipment labeling and outdoor labeling.
Compliance
RoHS Compliant, WEEE, EU REACH SVHC
†Compatible with the BMP™21-PLUS printer only.
Specifications
Specifications
Brown,White
6.4 m
0.375 in.
BMP™21 Series, ID PAL, LabPal Printers
21 ft.
9.53 mm