missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ High Traffic Series Heavy Roll

High Traffic Series Heavy Roll is a high absorbency mat for busy problem areas and industrial applications.

Manufacturer:  Brady™ HT550

 View more versions of this product

Catalog No. 19-150-280



Description

Description

  • Durable, high absorbency, dimpled roll
  • Used as pad, wipe, roll or soc
  • For use with oil and water-based fluids
  • Triple perfed lengthwise for 3 to 5 in. width dimensions and cross-wise every 18 in.
  • Universal class roll with 8gal/bale absorbency
  • Size: 38.1 x 127cm (15 x 50 in.)

RoHS Compatibility: Intended use of product is out of scope of the RoHS Directive

Walkways, Overspray Leaks and Drips, Workstations, Drums: Liquid Storage, Outfalls and Discharge Ponds

Specifications

Specifications

1/Cs.
Gray
1/Cs.
8gal/bale
High Traffic Series Heavy Roll
Documents