Brady™ Hazardous Materials Labels
Hazardous Materials Labels are black-on-yellow vinyl labels that provide workers with all of the chemical information they will need for hazardous materials they may encounter.
Manufacturer:  Brady™ BRD93477
Catalog No. 29-635-5219
Description
Description
  • The B-928 is a flexible vinyl film with a cold temperature applicable pressure sensitive adhesive
  • Service temperature range of -50 to 176°F
  • Average outdoor durability of 2 years
  • RoHS Compatibility Pass 2011/65/EU