Please login to your online account to display your discounted pricing

Brady™ GlobalMark™ Portable Label Maker

Makes customized labels on the spot

$26.23 - $141.36

Specifications

For Use With (Application) Ideal for any marking or identification
For Use With (Equipment) HandiMark™ portable label maker
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    

19-034720

 
brady™
BRD42007
Battery Charger Each for $62.07

19-034721

 
brady™
BRD42008
Spare Battery Pack Each for $141.36

19-034717

 
brady canada
BRD42004
PC Cable Kit Each for $26.23
Description & Specifications

Specifications

For Use With (Application) Ideal for any marking or identification
For Use With (Equipment) HandiMark™ portable label maker

Ideal for any marking or identification application, including pipe markers and valve tags; maintenance, equipment, instruction, warehouse, and safety labels; and laboratory applications. Can be used in the office, throughout the plant, or in the field.

  • Lightweight, hand-held label maker creates made-to-order labels quickly and easily, with no secondary operations
  • Contains a library of more than 100 symbols and pictograms
  • Operates in at least 12 resident languages
  • Automatic type sizing and layout eliminates guesswork
  • Type ranges in size from 0.75 to 1.75 in. (1.91 to 4.45cm)
  • Type can be underlined, bolded, italicized, condensed, expanded, printed vertically, and customized
  • Bar coding capability and alphanumeric sequencing with user-defined index
  • 10,000-byte memory lets users create and store bar codes and frequently used labels
  • Uses easy-to-load, industrial-strength label tapes and ribbons, available in a wide range of colors and designed that meet all color and regulatory requirements (order separately)

Dimensions (L x W x H): 12 × 3.75 × 4.5 in. (30.5 × 9.53 × 11.4cm). Runs on rechargeable battery or AC power supply. Optional accessories available.