Brady™ Globally Harmonized System (GHS) Oxidizing Picto Labels
For oxidizers, organic peroxides
Manufacturer:  Brady™ BRD118827
 View more versions of this product
Catalog No. 19-140-514
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
  • Legend: Oxidizing
  • Made of B-7569 Vinyl
  • Available in three sizes
Specifications
Specifications
Acrylic, Permanent
GHS, OSHA 29 CFR 1910.1200
0.62 in.
1.5cm
0.625 x 0.625 in.
-4 to 165°F
0.004 in.
Vinyl
39 Pack
Black, Red, White
Oxidizing
Oxidizers
3 Year(s)
1.58 x 1.58cm
-20 to 75°C
0.6 in.
Picto Label
Documents