Brady™ Globally Harmonized System (GHS) Explosive Picto Labels
For explosive, self-reactive, organic peroxides
Manufacturer:  Brady™ BRD118815
 View more versions of this product
Catalog No. 19-140-512
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.
  • Legend: Explosive
  • Made of Vinyl or Coated paper
  • Available in three sizes
Specifications
Specifications
Acrylic, Permanent
0.62 in.
1.57cm
-20 to 75°F
0.004 in.
Vinyl
39 Pack
Black, Red, White
Explosives, Self reactives, Organic Peroxides
3 Year(s)
-4 to 167°C
0.6 in.
Picto Label
Documents