Brady™ Globally Harmonized System (GHS) Corrosive Picto Labels
For Corrosives
Manufacturer:  Brady™ BRD118839
 View more versions of this product
Catalog No. 19-140-516
Description
Description
For Corrosives
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
  • Legend: Corrosive
  • Made of B-7569 Vinyl or Paper
  • Available in three sizes
Specifications
Specifications
Acrylic, Permanent
GHS, OSHA 29 CFR 1910.1200
0.62 in.
1.5cm
-4 to 167°F
0.004 in.
Vinyl
39 Pack
Black, Red, White
Corrosive
For pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification
3 Year(s)
-20 to 75°C
0.6 in.
Picto Label
Documents