Please login to your online account to display your discounted pricing

Brady™ FreezerBondz Thermal Transfer Printer Labels: Rectangular with Circles

Perform well in common laboratory environments such as freezing spray

$174.95 - $174.95

Specifications

Adhesive Type Acrylic
Adhesive Type Permanent
Color White
Product Type Laboratory Label
Finish Matte
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Height (English) Quantity Width (English) Price Quantity    

22-500-519

 
brady™
THT1634923
1 in. 0.37 in. 3000 Roll(s) 1 in. Each for $174.95
Description & Specifications

Specifications

Adhesive Type Acrylic
Adhesive Type Permanent
Color White
Product Type Laboratory Label
Finish Matte
For Use With (Application) Laboratory identification
Material FreezerBondz™ Polyester
Temperature Range (English) -320-266°F
Temperature Range (Metric) -160-130°C

Dual format: rectangular and circular label.

  • Excellent low temperature performance
  • Can be applied to frozen surfaces, including glass and polypropylene stored in liquid nitrogen
  • Offer excellent print smudge and solvent resistance (when used with the R6400 ribbon)

Laboratory Identification