missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Freezerbondz Polyester (B-490) LABXPERT™ Labels

Identification label for frozen surfaces including glass and polypropylene stored in liquid nitrogen

$70.07 - $77.82

Specifications

Color Black,White
Finish Matte
For Use With (Equipment) LABXPERT Printer
Material FreezerBondz™ Polyester
Coating Smooth
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Size (English) Price Quantity  
Catalog Number Mfr. No. Size (English) Price Quantity  
22-500-746 Brady™
BRD60908
1 x 1.750 in. Pack of 225 Each for $70.07
22-500-747 Brady™
BRD60909
0.6 x 1.8 in. Pack of 500 Each for $70.07
22-500-742 Brady™
X112490*
1.25 x 0.375 in. Pack of 500 Each for $77.82
Specifications

Description

  • Smooth surface label made of Freezerbondz Polyester
  • Moisture resistant label with cold-weather performance
  • Original Freezerbondz I label withstands extremely low temperatures
  • For LABEXPERT™ printer (also fits in IDXPERT™ printer)
  • Matte finish, black on white label
  • Environment: Autoclave, Freezer, Hot Water Baths (100°C), Liquid Nitrogen
  • Available in different sizes and printable areas
  • Printer compatibility: LABXPERT

Intended use of product is out of scope of the RoHS Directive

Description & Specifications

Specifications

Black,White
LABXPERT Printer
Smooth
Matte
FreezerBondz™ Polyester