missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Floor Safety Signs

Durable, highly visible Floor Safety Signs are designed to warn of hazardous areas, putting valuable safety information at the point of need.

Manufacturer:  Brady™ BRD97614

 View more versions of this product

Catalog No. 19-054-901



Description

Description

  • Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
  • B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
  • Excellent chemical, oil and water resistance
  • Minimum application temperature of 0°F (-18°C)
Specifications

Specifications

Abrasion Resistance, Method 5306 of U.S. Federal Test Method Std. No. 191A
White on Red
Vinyl
Permanent Acrylic
Designed to warn of hazardous areas, putting valuable safety information at the point of need
-40°F
Floor Safety Sign
-40°F to 158°F
Stop
1 Ea.
17 in.
158°F
0.01 in.
-40°C to 70°C
SDS
Documents
Product Certifications