missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Floor Safety Signs
Durable, highly visible Floor Safety Signs are designed to warn of hazardous areas, putting valuable safety information at the point of need.
Manufacturer: Brady™ BRD97610
Description
- Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
- B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
- Excellent chemical, oil and water resistance
- Minimum application temperature of 0°F (-18°C)
Specifications
Abrasion Resistance, Method 5306 of U.S. Federal Test Method Std. No. 191A | |
Black/Purple on White | |
Vinyl | |
Permanent Acrylic | |
Floor Mount | |
158°F | |
0.01 in. | |
-40°C to 70°C |
-40°F to 158°F | |
Keep Aisles Clear | |
1 Ea. | |
17 in. | |
Designed to warn of hazardous areas, putting valuable safety information at the point of need | |
-40°F | |
Floor Safety Sign |