missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Floor Safety Signs

Durable, highly visible Floor Safety Signs are designed to warn of hazardous areas, putting valuable safety information at the point of need.

Manufacturer:  Brady™ BRD97609

 View more versions of this product

Catalog No. 19-054-897



Description

Description

  • Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
  • B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
  • Excellent chemical, oil and water resistance
  • Minimum application temperature of 0°F (-18°C)
Specifications

Specifications

Abrasion Resistance, Method 5306 of U.S. Federal Test Method Std. No. 191A
Green on White
Vinyl
Permanent Acrylic
Floor Mount
158°F
0.01 in.
-40°C to 70°C
-40°F to 158°F
Eye Wash
1 Ea.
17 in.
Designed to warn of hazardous areas, putting valuable safety information at the point of need
-40°F
Floor Safety Sign
SDS
Documents
Product Certifications