Brady™ Fargo Production Status Tags
Fargo Production Status Self-Laminating tags can be personalized by writing on special instructions, hazards or warnings.
$34.23 - $34.23
Specifications
Certifications/Compliance ASTM D1593, ASTM D638
Material Polyester
Durability 3 to 5 Years
For Use With (Application) For marking on-the-spot warning of dangerous conditions
Height (English) 5.75 in.
View More Specs
Products 1
Catalog Number Mfr. No. Color Message Price Quantity  
Catalog Number Mfr. No. Color Message Price Quantity  
19-040-543 View Documents Brady™
BRD86457
Black on Orange Header: Warning Pack of 10 Each for $34.23
Description
Description
  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates and other special instructions
  • Seal and protect your written message with a clear adhesive overlay
  • Withstands moisture, grease and dirt
Specifications
Specifications
ASTM D1593, ASTM D638
3 to 5 Years
5.75 in.
10 nylon ties
-40° to 120°C
3 in.
Polyester
For marking on-the-spot warning of dangerous conditions
14.6cm
-40° to 248°F
0.36mm
7.62cm