missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Electrical Hazard Signs: WARNING - HIGH VOLTAGE

Identify electrical hazard and high voltage areas

Manufacturer:  Brady™ BRD69586

 View more versions of this product

Catalog No. 19-102-290



Description

Description

  • Available in two materials - plastic and fiberglass
  • Black on orange

Specifications

Specifications

ANSI Z535, OSHA 1910.145, PSTC-33,
Black on Orange
Fiberglass
9000 psi
Industrial, utility, commercial and institutional environments for both indoor and outdoor use
25.40 cm
-40°F
Electrical Hazard Sign
35.56 cm
-40°F to 193°F
Warning High Voltage
1 Ea.
Surface Mount
10 in.
193°F
0.1 in.
14 in.
-40°C to 90°C
SDS
Documents