missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Electrical Hazard Signs: Legend: DANGER - CABLE BURIED HERE
Identify electrical hazard and high voltage areas
Supplier: Brady™ 89098
Description
- Available in three materials - premium fiberglass, plastic and self sticking polyester
- Available in different widths and heights
- Black/red on white
Specifications
PSTC-33 | |
1 Ea. | |
7 in. | |
230°F | |
0.006 to 0.01 in. | |
25.40 cm | |
-40°F to 230°F | |
Electrical Hazard Sign |
Polyester | |
Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label | |
17.78 cm | |
-40°F | |
10 in. | |
-40°C to 110°C | |
DANGER Cable Buried Here | |
Red on White |