missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Electrical Hazard Signs: DANGER - HIGH VOLTAGE VOLTS
Identify electrical hazard and high voltage areas
$21.57 - $36.88
Specifications
Quantity | 1 Ea. |
---|---|
Mounting Type | Surface Mount |
Height (English) | 10 in. |
Height (Metric) | 25.40 cm |
Min. Temperature (English) | -40°F |
Catalog Number | Mfr. No. | Certifications/Compliance | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Price | Quantity | |||||
19035904
|
Brady™
25553 |
ASTM D 1593, OSHA 1910.145 |
Each for $21.57
|
|
|||||
19-035-906
|
Brady™
84904 |
ANSI Z535, OSHA 1910.145, PSTC-33, |
Each for $36.88
|
|
|||||
Description
- Available in two materials - plastic and polyester
- Available in combination of two different widths and heights
- Black/red on white
1 Use 1 in. numbers.
Specifications
1 Ea. | |
10 in. | |
-40°F | |
35.56 cm | |
Electrical Hazard Sign |
Surface Mount | |
25.40 cm | |
14 in. | |
DANGER High Voltage Volts |