Brady™ E-Z Check™ Plus GFI Circuit Tester
Checks for variety of electrical conditions and hazards
Supplier: Brady™ 65273
Description
Checks for following conditions:- Correct wiring, open ground
- Reverse polarity
- Open hot
- Open neutral
- Hot on neutral
- Hot and ground reversed with open hot
- Ground fault interruption
Specifications
Circuit Tester |