Brady™ “Do Not Operate” Lockout Tags
“Do Not Operate” Lockout Tags are perfect for marking on-the-spot warnings of dangerous conditions.
Manufacturer: Brady™ BRD66050
Description
- Rigid economy polyester and cardstock write-on tags meet OSHA Standard 1910.145 for accident prevention.
- Durable heavy duty write-on polyester (B-851) tags with brass grommets exceeds OSHA's 50 lb pull force requirement for tagout devices.
- Withstands moisture, grease and dirt.
- Paper cardstock (B-853) tags are recommended for indoor use only.
Specifications
OSHA 1910.145 | |
Danger Do Not Operate This lock/tag may only be removed by ETC, Back This energy source has been Locked Out! Unauthorized removal of This lock/tag may result in immediate discharge. Remarks | |
Marking on the spot warning of dangerous conditions | |
146 mm | |
−40° to +100°C | |
3 in. | |
Black/Red on White | |
Lockout Tag |
5.75 x 3 in. | |
3 to 5 Years | |
5.75 in. | |
−40° to +212°F | |
0.25 mm | |
76 mm | |
Polyester |