missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ “Danger: Hazardous Waste” Signs
For identifying equipment, rooms and materials that contain or are contaminated with hazardous agents
Supplier: Brady™ 85500
Description
- Bright colors and bold text provide high visibility to critical messages
- Sign reads DANGER HAZARDOUS WASTE
- Black/red on white
- Designed to withstand harsh industrial environments
- Available in different sizes and materials
B-401
- Plastic — high-impact polystyrene
- Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
Specifications
ANSI Z535, OSHA 1910.145 | |
1 Ea. | |
Identification of fire alarms, extinguishers and emergency exits | |
25.4 cm | |
-40°F | |
14 in. | |
-40°C to 110°C | |
DANGER Hazardous Waste | |
Red on White |
Polyester | |
Surface Mount | |
10 in. | |
230°F | |
0.006 to 0.01 in. | |
35.56 cm | |
-40°F to 230°F | |
Chemical and Hazardous Materials Sign |