missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ “Danger: Acid” Signs

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

Manufacturer:  Brady™ BRD84326

 View more versions of this product

Catalog No. 19-102-101



Description

Description

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “DANGER ACID”
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Specifications

    ANSI Z535, OSHA 1910.145
    Black/Red on White
    Polyester
    Surface Mount
    7 in.
    230°F
    0.006 to 0.01 in.
    10 in.
    -40°C to 110°C
    -40°F to 230°F
    DANGER Acid
    1 Ea.
    Identification of fire alarms, extinguishers and emergency exits
    17.7 cm
    -40°F
    Chemical and Hazardous Materials Sign
    25.4 cm
    SDS
    Documents