Please login to your online account to display your discounted pricing

Brady™ Conduit and Voltage Markers

$2.91 - $2.91

Specifications

Certifications/Compliance ASME (ANSI) ZG3.22
Adhesive Type Repositionable
Adhesive Type Rubber
Color Black
Color Orange
View More Specs
Products
Catalog Number Mfr. No. Description Display Text Height (Metric) Quantity Price Quantity    

19-015657

 
brady™
BRD44210
B 240 Volts 2.8cm 4 Card Each for $2.91

19-015669

 
brady™
BRD44260
B 277/480 Volts 2.8cm 4 Card Each for $2.91

19-015672

 
brady™
BRD44308
C 230 Volts 1.2cm 18 Card Each for $2.91

19-015673

 
brady™
BRD44315
C 480 Volts 1.2cm 18 Card Each for $2.91

19-015676

 
brady™
BRD44359
C 120/208 Volts 1.2cm 18 Card Each for $2.91

19-015675

 
brady™
BRD44331
C Three Phase 1.2cm 18 Card Each for $2.91

19-015650

 
brady™
BRD44115
A 480 Volts 5.7cm 1 Card Each for $2.91
Description & Specifications

Specifications

Certifications/Compliance ASME (ANSI) ZG3.22
Adhesive Type Repositionable
Adhesive Type Rubber
Color Black
Color Orange
Product Type Conduit and Voltage Marker Label
Finish Semi-Gloss
For Use With (Application) For wire, cable and general identification numbers, letters and conduit and voltage markers
Material Vinyl Cloth