Please login to your online account to display your discounted pricing

Brady™ Chemical and Hazardous Materials Signs: DANGER - FLAMMABLE NO MATCHES OR OPEN LIGHTS

Identify equipment, rooms and materials which contain or are contaminated with hazardous agents

$8.18 - $25.31

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145
Color Red, Black on White
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Message Flammable No Matches or Open Light
Type Chemical and hazardous materials sign
View More Specs
Products
Catalog Number Mfr. No. Height (English) Material Thickness Width (English) Price Quantity    

19-035-969

 
brady™
BRD85187
7 in. Polyester 0.006 to 0.01 in. 10 in. Each for $8.18

19-035-970

 
brady™
BRD85188
10 in. Polyester 0.006 to 0.01 in. 14 in. Each for $11.49

19-035-966

 
brady™
BRD71945
10 in. Fiberglass 0.1 in. 14 in. Each for $25.31
Description & Specifications

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145
Color Red, Black on White
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Message Flammable No Matches or Open Light
Type Chemical and hazardous materials sign
Quantity Each
Style Surface Mount

  • Available in three materials - plastic, fiberglass and polyester
  • Available in different widths and heights
  • Black/red on white