missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Chemical and Hazardous Materials Signs: DANGER - FLAMMABLE LIQUIDS
Identify equipment, rooms and materials which contain or are contaminated with hazardous agents
Manufacturer: Brady™ BRD72238
Description
- Available in three materials - plastic, fiberglass and polyester
- Available in various widths and heights
- Black/red on white
Specifications
ANSI Z535, OSHA 1910.145 | |
Each | |
9000psi | |
10 in. | |
193°F | |
-40°F | |
0.1 in. | |
14 in. |
Fiberglass | |
Surface Mount | |
Designed for use in industrial, utility, commercial and institutional environments | |
25.40cm | |
90°C | |
-40°C | |
Chemical and Hazardous Materials Sign | |
35.56cm |