Please login to your online account to display your discounted pricing

Brady™ “Caution: Safety Glasses Must Be Worn At All Times” Signs

Provides strong and noticeable reminder for your employees to protect themselves and maintain safe work atmosphere

$8.83 - $11.49

Specifications

Certifications/Compliance OSHA 1910.145, PSTC-33
Color Black on Yellow
For Use With (Application) Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label
Material Polyester
Message Caution Safety Glasses Must Be Worn At All Times
View More Specs
Products
Catalog Number Mfr. No. Height (English) Price Quantity    

19-036273

 
brady™
BRD84990
10 in. Each for $11.49

19-102154

 
brady™
BRD84989
7 in. Each for $8.83
Description & Specifications

Specifications

Certifications/Compliance OSHA 1910.145, PSTC-33
Color Black on Yellow
For Use With (Application) Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label
Material Polyester
Message Caution Safety Glasses Must Be Worn At All Times
Temperature Range (English) -40 to 230°F
Temperature Range (Metric) -40 to 110°C
Thickness 0.006 to 0.01 in.
Type Eye protection sign
Quantity Each
Style Surface Mount

  • Sign reads “Caution: Safety Glasses Must Be Worn At All Times” Signs
  • Available in range of size, materials and legends suitable for many different facilities and environments
  • Black on yellow