Please login to your online account to display your discounted pricing

Brady™ “Caution: Safety Belt And A Lifeline Required” Signs

For communicating required protective gear for confined spaces

$8.18 - $11.49

Specifications

Color Black on Yellow
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Message Safety Belt And A Lifeline Required
Type Confined space sign
Quantity Each
View More Specs
Products
Catalog Number Mfr. No. Width (English) Price Quantity    

19-035-788

 
brady™
BRD84558
10 in. Each for $8.18

19-035-789

 
brady™
BRD84559
14 in. Each for $11.49
Description & Specifications

Specifications

Color Black on Yellow
For Use With (Application) Designed for use in industrial, utility, commercial and institutional environments
Message Safety Belt And A Lifeline Required
Type Confined space sign
Quantity Each
Style Surface Mount

  • Bright colors and bold text provide high visibility to critical messages
  • Legend: SAFETY BELT AND A LIFELINE REQUIRED
  • Header: CAUTION
  • Black on Yellow
  • Available in different sizes and materials

B-302

  • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
  • B-401
    • Plastic — high-impact polystyrene