Brady™ BSP™31 Label Attachment System
For histology and anatomical pathology specimen labeling
Manufacturer:  Brady™ BSP31
 View more versions of this product
Catalog No. 29-636-0748
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
For histology and anatomical pathology specimen labeling
The BSP31 Label Attachment System gives labs the ability to permanently attach labels to tissue cassettes, resulting in more accurate specimen tracking and sample management.
  • Pre-process identification for tissue samples allowing traceability throughout the laboratory processes and storage
  • Provides greater efficiency; addresses market need for labeling small volumes of tissue cassettes
  • Reduces the chance of identification errors and reduces the liability
  • Backup system for high volume laboratories
  • Must be used exclusively with Brady tissue cassette label material and Brady thermal transfer printer ribbon
Specifications
Specifications
Red
6.51 in.
16.53 cm
68 to 80°F
13 lb.
6.45 in.
Label Attachment System
UL, CSA, EN, IEC 60950-1, EMC,2004/108/EEC, RoHS, 2002/95/EC, ISO 9614-1
For histology and anatomical pathology specimen labeling
Attachment system only
18 to 27°C
5.9 kg
Label
Each
Documents