missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ BSP™31 Label Attachment System

For histology and anatomical pathology specimen labeling

$4,436.32 - $5,595.31

Specifications

Certifications/Compliance UL, CSA, EN, IEC 60950-1, EMC,2004/108/EEC, RoHS, 2002/95/EC, ISO 9614-1
Color Red
For Use With (Application) For histology and anatomical pathology specimen labeling
Height (English) 6.51 in.
Height (Metric) 16.53cm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
29-636-0748 Brady™
BSP31
Attachment system only Each for $4,436.32
29-636-0749 Brady Canada
BSP31BASIC
System w/3-year basic program Each for $5,595.31
Specifications

Description

The BSP31 Label Attachment System gives labs the ability to permanently attach labels to tissue cassettes, resulting in more accurate specimen tracking and sample management.

  • Pre-process identification for tissue samples allowing traceability throughout the laboratory processes and storage
  • Provides greater efficiency; addresses market need for labeling small volumes of tissue cassettes
  • Reduces the chance of identification errors and reduces the liability
  • Backup system for high volume laboratories
  • Must be used exclusively with Brady tissue cassette label material and Brady thermal transfer printer ribbon

Description & Specifications

Specifications

UL, CSA, EN, IEC 60950-1, EMC,2004/108/EEC, RoHS, 2002/95/EC, ISO 9614-1
For histology and anatomical pathology specimen labeling
16.53cm
6.45 in.
Red
6.51 in.
18 to 27°C