Please login to your online account to display your discounted pricing

Brady™ BMP™53 Label Maker Printer

Compact, versatile portable printer with built-in Bluetooth™ technology for wireless printing

$505.56 - $832.06

Specifications

Connector Types USB, Bluetooth and optional Ethernet, Wi-Fi
Display Backlit LCD
For Use With (Application) Audio or visual labeling, data communications labeling, electrical labeling, facility, safety and maintenance identification, laboratory identification, product identification
Resolution 300 dpi
Warranty 1 Year(s)
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

19-155-102

 
brady™
BMP53KITLMK
BMP™53 printer, USB cable, 1 MC-1500-595-WT-BK label cartridge, hardcase, quick start guide, AC adapter and Labelmark™ 5 standard software and Labelmark™ Each Each for $593.50

19-155-101

 
brady™
BMP53
BMP™53 printer, USB cable, MC-1500-595-WT-BK label cartridge, hardcase, quick start guide and AC adapter/battery charger Each Each for $505.56

19-018-018

 
brady™
BMP53KITLS
BMP™53 printer, USB cable, hardcase, quick start guide, AC adapter/battery charger, Labelmark™ standard software and M-156-492, M-81-48, M-120-492, M-103-498, MC-1500-595-WT-BK label cartridges 1 Kit Each for $832.06
Description & Specifications

Specifications

Connector Types USB, Bluetooth and optional Ethernet, Wi-Fi
Display Backlit LCD
For Use With (Application) Audio or visual labeling, data communications labeling, electrical labeling, facility, safety and maintenance identification, laboratory identification, product identification
Resolution 300 dpi
Warranty 1 Year(s)
Weight (English) 2.7 lb.
Weight (Metric) 1.22kg

  • Prints up to 1.5 in. wide labels
  • 300 dpi print head
  • Print speed: 1 in. per second
  • Automatic label formatting
  • Durable built-in cutter
  • USB PC Connectivity: create more advanced labels using LabelMark™ Software with barcode symbologies, time/date stamps, imported graphics, automatic data serialization and more
  • Battery options for portability: printer runs on AA batteries, the optional rechargeable lithium-ion battery or AC power supply
Hassle-Free Printer Cartridges:
  • Ribbon and label materials in one cartridge
  • Install or remove ribbon and material in one easy step
  • No need to worry about having the right ribbon for your materials
  • Ribbon and material runs out at the same time (less waste)
  • Top-loading cartridge with “no jam” technology that snaps into place