Please login to your online account to display your discounted pricing

Brady™ BMP™50 Series Printable Repositionable Vinyl Cloth Labels

Designed for use with various BMP™50 series label printers. Brady™ BMP™50 Series Printable Repositionable Vinyl Cloth Labels offer a repositionable adhesive and are intended for use in wire and cable marking, component or equipment identification, barcoding, and other general identification.

$34.50 - $59.42

Specifications

Material Vinyl Cloth (B-498)
Adhesive Type Repositionable
Adhesive Type Rubber
Color Black
Color White
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Includes Size (English) Price Quantity    

19-155-149

 
brady™
M131498
BMP™41, BMP™51, and BMP™53 printers 180 labels 0.5 x 1 in. Each for $59.42

19-155-151

 
brady™
M103498
BMP™51 and BMP™53 printer 160 labels 1 x 1.25 in. Each for $34.50
Description & Specifications

Specifications

Material Vinyl Cloth (B-498)
Adhesive Type Repositionable
Adhesive Type Rubber
Color Black
Color White
Certifications/Compliance RoHS, UL
Display Text None
Finish Semi-Gloss
Format Cartridge
For Use With (Application) Barcoding, component ID, equipment ID, general purpose ID, wire and cable marking
Life Span 2 Years
Print Technology Thermal transfer
Temperature Range (English) -40 to 176°F
Temperature Range (Metric) -40 to 80°C

  • Adhesive and cloth backing offers excellent holding power, yet allow for clean removal and repositioning
  • Stick to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • All-in-one cartridge includes label supply and R6200 Series ribbon