Brady™ BMP™ 21 ID Pal™ Label Cartridges: Self-Laminating Vinyl
Self-laminating vinyl thermal transfer labels
Manufacturer:  Brady™ M211000427
 View more versions of this product
Catalog No. 19-102-710
Description
Description
  • Sticks well to both smooth and textured surfaces
  • Allows you to label cables and wires with a simple wrapping function
  • Provides excellent smudge resistant text
Specifications
Specifications
Label Cartridge
Black,Translucent,White
Self-laminating Vinyl tape for BMP21-PLUS; BMP21-LAB; BMP21; IDPAL; LABPAL
BMP™21
4.30 m
0.102 mm
Vinyl
Cartridge, Label
14 ft.
25.40 mm
Acrylic,Permanent
UL
Communications cables, General identification, Laboratory identification, Panel identification, Voice or data identification, Wire and cable marking
4.2 m
-70°C to +70°C
9.53 mm
1 Roll
Matte
1 in.