Brady™ BMP™ 21 ID Pal™ Label Cartridges: Nylon Cloth for Wire, Cable and Panels
For wire, cable, faceplates and patch panels
Manufacturer:  Brady™ M21375499
 View more versions of this product
Catalog No. 19-102-720
Description
Description
For wire, cable, faceplates and patch panels
  • Flexible nylon cloth material
  • Adheres best to highly textured, low surface energy, powder-coated and smooth surfaces
Specifications
Specifications
Label Cartridge
Black,White
Nylon Cloth tape for BMP21-PLUS; BMP21; IDPAL; LABPAL
BMP™21, ID PAL, LABPAL
1
-40°C to +90°C
Nylon Cloth
Cartridge, Label
16 ft.
9.53 mm
Acrylic,Permanent
Halogen-free (DIN VDE 0472/parts 815), UL
Communications cables, General identification, Laboratory identification, Panel identification, Voice or data identification, Wire and cable marking
4.8 cm
4.88 m
0.165 mm
1 Roll
Matte
0.3 in.