Brady™ BLS 850 Laminator
Portable desktop laminator works without electricity, heat, or chemicals
$203.88 - $203.88
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
19-034787 Brady™
BRD20585
Each for $203.88
Specifications
Description
  • Ideal for office use or industrial applications
  • Integral cut-off blade at roller exit
  • Simply insert the appropriate cartridge to laminate one or both sides of a document or to apply laminate to one side and adhesive to the other
  • Laminates documents up to 8.5 in. × 100 ft. (21.6cm × 30.5m)
  • Cartridges use industrial-grade overlaminate constructed of tough polyester for durability even in harsh environments
Order Info
Cartridges sold separately.