Brady™ “Biohazard” with Pictogram Signs
Identify equipment, rooms and materials that contain or are contaminated with hazardous agents
Manufacturer:  Brady™ BRD85553
 View more versions of this product
Catalog No. 29-664-064
Description
Description
Bright colors and bold text provide high visibility to critical messages.
  • Biohazard marking is required by safety code 1910.1030(g)(1)
  • Designed to withstand harsh industrial environments
  • Black on orange
  • B-485 self-sticking polyester — polyester film with sensitive acrylic
Compliance
Meets OSHA Standards 1910.145(e)(4) and 1910.1030(g)(1)
Specifications
Specifications
OSHA
Polyester
7 x 10 in.
7 in.
50°C
-40°F
0.01 in.
10 in.
Black on Orange
Each
Used to identify the actual or potential presence of a biologically hazardous material
122°F
Biohazard
-40°C
Biohazard sign
Documents