Brady™ Barricade Tapes
This durable, lightweight Barricade Tape is made of non-adhesive, glossy B-912 polyethylene for on-the-spot warnings of potential danger.
Manufacturer:  Brady™ BRD91468
Catalog No. 29-635-7912
Description
Description
  • Non-adhesive Polyethylene (B-912) is a glossy plastic that's ideal for temporary outdoor or permanent indoor use
  • Lightweight tape can be tied, stapled or nailed to posts, fences or metal barricades
  • Visual barricading provides fast on-the-spot warning of potential danger
  • B-912 polyethylene barricade tapes from Brady™ are available in variety of legends to fit your specific needs (each roll/legend is sold separately)
Specifications
Specifications
OSHA 1910.144
Authorized Personnel Only
Black,Yellow
Barricade Tape
Temporary outdoor or permanent indoor application
0.003 in.
7.6 cm
200 ft. x 3 in. (61m x 7.6 cm)
Safety
HDPE
200 ft.
61 m
3 in.
Documents