Brady™ Accident Prevention Tags with Same Fronts and Backs
Feature same message on tag front and back
Manufacturer:  Brady™ BRD86438
Catalog No. 29-635-8299
Description
Description
  • Feature two chamfered corners and a 3/8 in. grommeted hole
  • Textured write-on surface allows for authorized signatures, dates, and other special instructions
  • Chemical-resistant, plastic Dura-Tags are reusable and designed for harsh plant environments; come with nylon fasteners
  • Chemical-resistant economy polyester tags withstand moisture, grease and dirt; come with nylon fasteners
  • Economical, yet durable cardstock tags come with string fasteners
Compliance
Meet OSHA 1910.145 for accident prevention; Dura-Tag and polyester tags also meet OSHA 50 lb. pull requirement.
Specifications
Specifications
OSHA
0.93cm
Inspected Record Date/By/Accepted/Rejected
Up to 2 Years
5.75 in.
10 nylon fasteners
-40° to 80°C
3 in.
0.37 in.
Dura-Tag
For lockout and tagout identification, polyester laminate protects the graphics, wipes clean, and withstands water, grease, and extreme temperatures, heavy duty construction prevents accidental removal
3.9 Gardner
14.6cm
0.02 to 0.025 in.
-40° to 175°F
7.6cm