Brady™ Accident Prevention Tags with Same Fronts and Backs
Feature same message on tag front and back
Supplier: Brady™ 86535
Description
- Feature two chamfered corners and a 3/8 in. grommeted hole
- Textured write-on surface allows for authorized signatures, dates, and other special instructions
- Chemical-resistant, plastic Dura-Tags are reusable and designed for harsh plant environments; come with nylon fasteners
- Chemical-resistant economy polyester tags withstand moisture, grease and dirt; come with nylon fasteners
- Economical, yet durable cardstock tags come with string fasteners
Compliance
Meet OSHA 1910.145 for accident prevention; Dura-Tag and polyester tags also meet OSHA 50 lb. pull requirement.
Specifications
OSHA | |
Polyester | |
For marking on-the-spot warning of dangerous conditions | |
5.75 in. | |
10 nylon ties | |
-40°C to 100°C | |
3 in. | |
0.93 cm |
0.37 in. | |
Maintenance Hold Tag Maintenance Needed In Progress | |
3 to 5 Years | |
14.6cm | |
0.01 in. | |
-40°F to 212°F | |
7.6 cm |