Brady™ Accident Prevention Tags with Different Fronts and Backs
With different fronts and backs
Supplier: Brady™ 86694
Description
- Feature two chamfered corners and a 3/8 in. grommeted hole
- Textured write-on surface allows for authorized signatures, dates, and other special instructions
- Chemical-resistant, plastic Dura-Tags are reusable and designed for harsh plant environments; come with nylon fasteners
- Chemical-resistant economy polyester tags withstand moisture, grease and dirt; come with nylon fasteners
- Economical, yet durable cardstock tags come with string fasteners
Compliance
Meet OSHA 1910.145 for accident prevention; Dura-Tag and polyester tags also meet OSHA 50 lb. pull requirement.
Specifications
ASTM | |
0.37 in. | |
DANGER Stand Clear Automatic Or Remote Controlled Equipment Do Not Remove This Tag | |
7 in. | |
100 string fasteners | |
-40°C to 90°C | |
4 in. | |
0.93 cm |
Black, Red, White | |
Cardstock | |
Used for temporary tagging in a non harsh environment | |
17.7cm | |
0.01 in. | |
-40°F to 194°F | |
10.1 cm |