Brady™ 2-Gallon Spill Response Plus Retail Spill Kit
Compact size for convenient handling
Manufacturer:  Brady™ SKHSRP
 View more versions of this product
Catalog No. 19-140-746
Includes: 2 gal. bucket, 20 pads (7.5 x 10. in. ), 1 disposal bag, tie, 1 pair of nitrile gloves, 1 pair of goggles and instruction sheet
Description
Description
  • Highly absorbent, durable and low linting
  • Suitable for hospitality, retail, restaurants or anywhere that requires small pad
  • Absorbs up to 4.5 gal./kit
  • Dia. x H: 24.7 x 26.6cm (9.75 x 10.5 in.)
  • Available types: Universal, oil only and chemical
Specifications
Specifications
2 gal. container, 20 pads, 1 bag, tie, pair of gloves, goggles
24.7 x 26.6cm (9.75 x 10.5 in.)
Spill Kit, Chemical
Documents