Please login to your online account to display your discounted pricing

Alfa Aesar™ BQ-3020

For cellular and molecular biology applications

Specifications

Product Type BQ-3020
Molecular Weight 2006.32
Storage Requirements -30° to -10°C
Sequence N-Acetyl-Leu-Met-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66660EXD

 
alfa aesar™
J66660EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66660MCR

 
alfa aesar™
J66660MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type BQ-3020
Molecular Weight 2006.32
Storage Requirements -30° to -10°C
Sequence N-Acetyl-Leu-Met-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp
Format Solid
Regulatory Status RUO
Purification Method HPLC