Please login to your online account to display your discounted pricing

Alfa Aesar™ BQ-3020


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66660MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66660MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1mg
Format Solid
Sequence N-Acetyl-Leu-Met-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp
Molecular Weight 2006.32
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type BQ-3020
Purification Method HPLC