Please login to your online account to display your discounted pricing

Tocris™ BPIPP

Non-competitive GC inhibitor. Also AC inhibitor

$252.39 - $1,050.42

Specifications

Product Type BPIPP
Inhibitors BPIPP
Molecular Weight 450.28
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

363510R

 
tocris™
363510
10mg Each for $252.39

363550R

 
tocris™
363550
50mg Each for $1,050.42
Description & Specifications

Specifications

Product Type BPIPP
Inhibitors BPIPP
Molecular Weight 450.28
Purity >99

Non-competitive guanylyl cyclase (GC) and adenylyl cyclase (AC) inhibitor. Downregulates cAMP and cGMP synthesis, and suppresses cGMP accumulation in a variety of cell lines (IC50 = 3.4 - 11.2μM). Inhibits GC-stimulated Cl- transport in vitro and suppresses toxin-induced intestinal fluid accumulation in vivo.

  • Chemical Name: 5-(3-Bromophenyl)-5,11-dihydro-1,3-dimethyl-1H-indeno[2',1':5,6]pyrido[2,3-d]pyrimidine-2,4,6(3H)-trione
  • Formula: C22H16BrN3O3
  • CAS Number: 325746-94-9
  • Solubility: Soluble to 2mM in DMSO
  • Purity: >99