missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BP-1 Rat anti-Mouse, PerCP-eFluor 710, Clone: 6C3, eBioscience™

Rat Monoclonal Antibody

$201.55 - $520.55

Specifications

Antigen BP-1
Clone 6C3
Host Species Rat
Gene Alias Ly-51
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123859 Life Technologies
LS46589180
25μg Each for $201.55
501123860 Life Technologies
LS46589182
100μg Each for $520.55
Specifications

Description

The 6C3 monoclonal antibody reacts with mouse BP-1, also known as Ly-51 and 6C3 antigen. BP-1 is a 140kDa type II transmembrane protein that is present on the surface as a disulfide-linked homodimer. This molecule is an enzyme with aminopeptidase A (APA) activity and is expressed by bone marrow stromal cells, thymic cortical epithelial cells and at early stages of mouse B cell development, on pre-B cells. The low level of BP-1 expression on pre-B cells can be upregulated in the presence of IL-7.
Description & Specifications

Specifications

BP-1
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
6C3
Ly-51
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL