missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BP-1 Rat anti-Mouse, FITC, Clone: 6C3, eBioscience™

Rat Monoclonal Antibody

$117.48 - $209.88

Specifications

Antigen BP-1
Clone 6C3
Host Species Rat
Gene Alias BP1, Ly-51, Ly51
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509896 Life Technologies
LS11589181
50μg Each for $117.48
509897 Life Technologies
LS11589182
100μg Each for $209.88
Specifications

Description

The 6C3 monoclonal antibody reacts with mouse BP-1, also known as Ly-51 and 6C3 antigen. BP-1 is a 140kDa type II transmembrane protein that is present on the surface as a disulfide-linked homodimer. This molecule is an enzyme with aminopeptidase A (APA) activity and is expressed by bone marrow stromal cells, thymic cortical epithelial cells and at early stages of mouse B cell development, on pre-B cells. The low level of BP-1 expression on pre-B cells can be upregulated in the presence of IL-7.
Description & Specifications

Specifications

BP-1
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
6C3
BP1, Ly-51, Ly51
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL