PIP™ Bouton™ Recon™ Rimless Safety Glasses
Trendy rimless design offering ultimate comfort with non-slip bridge and end tips.
Manufacturer:  PIP™ EP250320226
 View more versions of this product
Catalog No. 19153363
Description
Description
  • Lightweight, rimless design provides all day user comfort
  • Flexible bayonet temples contain rubber pads for a secure, non-slip fit
  • Adjustable rubber nosepiece provides a comfortable fit
  • Indoor/Outdoor Blue Lens is a proprietary engineered lens that combines a slight yellow tint and a mirror coating, working in tandem to alleviate and help to prevent digital eye strain
  • The light yellow tint blocks the harsh blue light emitted from electronic screens and the slight mirror coating on the outside reduces glare from overhead lighting
  • Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
  • Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
Anti-fog,Scratch-resistant
Safety glasses
Anti-fog, Scratch-resistant
Blue,Indoor/Outdoor
Translucent Brown
Polycarbonate