Please login to your online account to display your discounted pricing

Alfa Aesar™ Bourgeonal, 99%


Quantity 5mg

Manufacturer: alfa aesar™  J67041LB0

 View more versions of this product

Catalog No. AAJ67041LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.29
Chemical Name or Material Bourgeonal
Physical Form Oil
Assay 99%
CAS 18127-01-0
Synonym 4-tert-Butylbenzenepropionaldehyde
Quantity 5mg
RTECS Number DA8100070
TSCA TSCA
Refractive Index 1.5100
MDL Number MFCD00169819
Color Colorless to pale yellow
Sensitivity Air Sensitive
Density 0.959g/mL
Storage Note 1 -30° to -10°C

Specifications

Chemical Name or Material Bourgeonal
CAS 18127-01-0
Formula Weight 190.29
MDL Number MFCD00169819
Synonym 4-tert-Butylbenzenepropionaldehyde
Accessories

-30° to -10°C

SDS & Safety Documents