Please login to your online account to display your discounted pricing

Alfa Aesar™ Bourgeonal, 99%

Chemical Identifiers

Chemical Name or Material Bourgeonal
CAS 18127-01-0
Formula Weight 190.29
MDL Number MFCD00169819
Synonym 4-tert-Butylbenzenepropionaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67041LB0

  SDS
alfa aesar™
J67041LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.29
Chemical Name or Material Bourgeonal
CAS 18127-01-0
Assay 99%
Quantity 5mg
Synonym 4-tert-Butylbenzenepropionaldehyde
RTECS Number DA8100070
Physical Form Oil
Color Colorless to pale yellow
Density 0.959g/mL
Storage Note 1 -30° to -10°C
Sensitivity Air Sensitive
Refractive Index 1.5100
TSCA TSCA
MDL Number MFCD00169819
Safety and Handling

-30° to -10°C