Please login to your online account to display your discounted pricing

Bouin's Fluid, Tissue Fixative (Picric Acid, Formaldehyde, Acetic Acid), LabChem™


Manufacturer: lab chem  LC117901

 View more versions of this product

Catalog No. LC117901

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 30.05
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid breathing mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
Contaminated clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container in accordance with local, state, and federal regulations.
CAS 50-00-0
Identification Passes Test
CAS Max % 83.3
CAS Max % 1.3
Decomposition Information Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Solubility Soluble in water
CAS 67-56-1
Recommended Storage Room Temperature
CAS 64-19-7
CAS Max % 4.8
UN Number UN3265
Chemical Name or Material Bouin's Fluid
Physical Form Liquid
CAS Max % 3.6
CAS Max % 7
Health Hazard 1 Danger
CAS 7732-18-5
Packaging Poly Bottle
CAS 88-89-1
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause cancer.
Causes damage to organs.
Toxic to aquatic life.
Color Clear, yellow
Linear Formula HCHO
Name Note Tissue Fixative (Picric Acid, Formaldehyde, Acetic Acid)

Specifications

Chemical Name or Material Bouin's Fluid
CAS 7732-18-5
CAS Max % 83.3
CAS 50-00-0
CAS Max % 7
CAS 64-19-7
CAS Max % 4.8
CAS 67-56-1
CAS Max % 3.6
CAS 88-89-1
CAS Max % 1.3
Name Note Tissue Fixative (Picric Acid, Formaldehyde, Acetic Acid)
Linear Formula HCHO
Formula Weight 30.05
UN Number UN3265
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid breathing mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
Contaminated clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container in accordance with local, state, and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause cancer.
Causes damage to organs.
Toxic to aquatic life.

Room Temperature