Please login to your online account to display your discounted pricing

Bouin's Fluid, Tissue Fixative (Picric Acid, Formaldehyde, Acetic Acid), LabChem™


Manufacturer: lab chem  LC117904

 View more versions of this product

Catalog No. LC117904

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 30.05
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause cancer.
Causes damage to organs.
Toxic to aquatic life.
Decomposition Information Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Linear Formula HCHO
CAS Max % 83.3
CAS Max % 7
CAS 88-89-1
Chemical Name or Material Bouin's Fluid
Recommended Storage Room Temperature
CAS 7732-18-5
UN Number UN3265
CAS 64-19-7
Name Note Tissue Fixative (Picric Acid, Formaldehyde, Acetic Acid)
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid breathing mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
Contaminated clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container in accordance with local, state, and federal regulations.
Solubility Soluble in water
Health Hazard 1 Danger
CAS Max % 3.6
CAS Max % 1.3
CAS Max % 4.8
Color Clear, yellow
Packaging Poly Bottle
CAS 50-00-0
Identification Passes Test
CAS 67-56-1
Physical Form Liquid

Specifications

Chemical Name or Material Bouin's Fluid
CAS 7732-18-5
CAS Max % 83.3
CAS 50-00-0
CAS Max % 7
CAS 64-19-7
CAS Max % 4.8
CAS 67-56-1
CAS Max % 3.6
CAS 88-89-1
CAS Max % 1.3
Name Note Tissue Fixative (Picric Acid, Formaldehyde, Acetic Acid)
Linear Formula HCHO
Formula Weight 30.05
UN Number UN3265
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid breathing mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
Contaminated clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container in accordance with local, state, and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause cancer.
Causes damage to organs.
Toxic to aquatic life.

Room Temperature