Please login to your online account to display your discounted pricing

Bouin's Fluid, Tissue Fixative (Picric Acid, Formaldehyde, Acetic Acid), LabChem™


Quantity 1L

Manufacturer: lab chem  LC117902

 View more versions of this product

Catalog No. LC117902

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 30.05
Chemical Name or Material Bouin's Fluid
CAS Max % 4.8
CAS 67-56-1
CAS Max % 3.6
CAS 88-89-1
CAS Max % 1.3
Identification Passes Test
Name Note Tissue Fixative (Picric Acid, Formaldehyde, Acetic Acid)
Physical Form Liquid
Packaging Poly Bottle
Linear Formula HCHO
CAS 7732-18-5
Solubility Soluble in water
UN Number UN3265
Health Hazard 1 Danger
Decomposition Information Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Color Clear, yellow
CAS Max % 83.3
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause cancer.
Causes damage to organs.
Toxic to aquatic life.
CAS 50-00-0
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid breathing mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
Contaminated clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container in accordance with local, state, and federal regulations.
Quantity 1L
CAS Max % 7
Recommended Storage Room Temperature
CAS 64-19-7

Specifications

Chemical Name or Material Bouin's Fluid
CAS 7732-18-5
CAS Max % 83.3
CAS 50-00-0
CAS Max % 7
CAS 64-19-7
CAS Max % 4.8
CAS 67-56-1
CAS Max % 3.6
CAS 88-89-1
CAS Max % 1.3
Name Note Tissue Fixative (Picric Acid, Formaldehyde, Acetic Acid)
Linear Formula HCHO
Formula Weight 30.05
UN Number UN3265
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid breathing mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
Contaminated clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container in accordance with local, state, and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause cancer.
Causes damage to organs.
Toxic to aquatic life.

Room Temperature