Please login to your online account to display your discounted pricing

Bouin's Fixative Solution, Ricca Chemical


Bouin's Fixative Solution; Picric acid, formalin, acetic acid mixture; Ricca Chemical

Quantity 1L
Packaging Poly Bottle

Manufacturer: ricca chemical company  RI112032

 View more versions of this product

Catalog No. 112032

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1L
Chemical Name or Material Bouin's Fixative Solution
CAS Min % 0.87
Packaging Poly Bottle
CAS 50-00-0
CAS Min % 24.33
CAS Max % 25.33
CAS 64-19-7
CAS Max % 4.96
CAS 88-89-1
CAS Max % 0.91
CAS 7732-18-5
CAS Min % 68.0
CAS Max % 71.0
CAS Min % 4.76

Specifications

Chemical Name or Material Bouin's Fixative Solution
CAS 7732-18-5
CAS Min % 68.0
CAS Max % 71.0
CAS 50-00-0
CAS Min % 24.33
CAS Max % 25.33
CAS 64-19-7
CAS Min % 4.76
CAS Max % 4.96
CAS 88-89-1
CAS Min % 0.87
CAS Max % 0.91